Isatin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2,3-Indolinedione, Indoline-2,3-dione, 1H-Indole-2,3-dione

M-Clarity™ quality level = MQ200

8.20709.0005 8.20709.0250
EM8.20709.0005CS 12044 USD
EM8.20709.0005 EM8.20709.0250
Isatin for synthesis, Sigma-Aldrich®
Isatin
Formula: C₈H₅NO₂
MW: 147.13 g/mol
MDL Number: MFCD00005718
CAS Number: 91-56-5

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Melting Point 199 - 204 °C
Molar Mass 147.13 g/mol
Bulk Density 600 kg/m3
Flash Point 200 °C
pH 7 (1.9 g/l, H2O, 20 °C)
Ignition Point 220 °C
Hill Formula C8H5NO2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR