5-(Hydroxymethyl)-2-furaldehyde for synthesis, Sigma-Aldrich®
Supplier: MERCK, S.A. DE C.V. SV
|
Warning
|
M-Clarity™ quality level = MQ200
8.20678.0001
8.20678.0005
EM8.20678.0001CS
4654
USD
EM8.20678.0001
EM8.20678.0005
5-(Hydroxymethyl)-2-furaldehyde for synthesis, Sigma-Aldrich®
5-(Hydroxymethyl)-2-furaldehyde
Formula:
C₆H₆O₃ |
MDL Number:
MFCD00003234 CAS Number: 67-47-0 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 27 - 30 °C |
Molar Mass | 126.11 g/mol |
Boiling Point | 154 - 155 °C (16 hPa) |
Flash Point | 79 °C |
Refractive Index | 1.5627 (20 °C, 589 nm) |
Synonyms | 5-(Hydroxymethyl)furfurol |
Hill Formula | C6H6O3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...