Cacodylic acid sodium salt trihydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20670.0025 8.20670.0100 8.20670.0250
EM8.20670.0025CS 8752 USD
EM8.20670.0025 EM8.20670.0100 EM8.20670.0250
Cacodylic acid sodium salt trihydrate for synthesis, Sigma-Aldrich®
Cacodylic acid sodium salt trihydrate
Formula: (CH₃)₂As(O)ONa·3H₂O
MDL Number: MFCD00149079
CAS Number: 6131-99-3
UN: 1688
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 2000 g/l (20 °C)
Melting Point 60 °C
Molar Mass 214.03 g/mol
Bulk Density 870 kg/m3
pH 9.0 - 9.5 (100 g/l, H2O, 20 °C)
Synonyms Dimethylarsinic acid sodium salt, Cacodylic acid sodium salt
Hill Formula C2H6AsNaO2 * 3 H2O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR