1,4-Butanesultone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1,4-Butane sultone

M-Clarity™ quality level = MQ200

8.20665.0050 8.20665.0250
EM8.20665.0050CS 8851 USD
EM8.20665.0050 EM8.20665.0250
1,4-Butanesultone for synthesis, Sigma-Aldrich®
1,4-Butanesultone
Formula: C₄H₈O₃S
MW: 136.17
Boiling Pt: 153
Density: 1.331
MDL Number: MFCD00006584
CAS Number: 1633-83-6

Order Now

Specification Test Results

Solubility 54 g/l (20 °C) (decomposition)
Melting Point 12.5 - 15.0 °C
Molar Mass 136.16 g/mol
Boiling Point 153 °C (19 hPa)
Flash Point 154 °C
Refractive Index 1.462 (25 °C, 589 nm)
Density 1.34 g/cm3 (20 °C)
Synonyms 4-Hydroxy-1-butanesulfonic acid δ-sultone
Hill Formula C4H8O3S


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR