Salicyl alcohol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20662.0050 8.20662.0250
EM8.20662.0050CS 1536 USD
EM8.20662.0050 EM8.20662.0250
Salicyl alcohol for synthesis, Sigma-Aldrich®
Salicyl alcohol
Formula: C₇H₈O₂
MDL Number: MFCD00004617
CAS Number: 90-01-7

Order Now

Specification Test Results

Solubility 63 g/l (20 °C)
Melting Point 83 - 86 °C
Molar Mass 124.14 g/mol
Synonyms Saligenine, Salicyl alcohol
Hill Formula C7H8O2
Chemical Formula 2-(OH)C6H4CH2OH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR