4-Hydroxybenzophenone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
(4-Hydroxyphenyl)(phenyl)methanone, p-Hydroxybenzophenone
M-Clarity™ quality level = MQ200
8.20661.0025
8.20661.0100
EM8.20661.0025CS
5354
USD
EM8.20661.0025
EM8.20661.0100
4-Hydroxybenzophenone for synthesis, Sigma-Aldrich®
4-Hydroxybenzophenone
Formula:
C₁₃H₁₀O₂ |
MDL Number:
MFCD00002355 CAS Number: 1137-42-4 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 131 - 133 °C |
Molar Mass | 198.22 g/mol |
Boiling Point | 260 - 262 °C (32 hPa) |
Synonyms | p-Benzoylphenol |
Hill Formula | C13H10O2 |
Chemical Formula | 4-(OH)C6H4COC6H5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...