Dihexylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20465.0100 8.20465.2500
EM8.20465.0100CS 1418 USD
EM8.20465.0100 EM8.20465.2500
Dihexylamine for synthesis, Sigma-Aldrich®
Dihexylamine
Formula: C₁₂H₂₇N
MW: 185.35 g/mol
MDL Number: MFCD00009521
CAS Number: 143-16-8
UN: 2927
ADR: 6.1,I

Order Now

Specification Test Results

Solubility 0.3 g/l (25 °C)
Melting Point 3 °C
Molar Mass 185.35 g/mol
Boiling Point 236 °C (1013 mbar)
Vapor Pressure 0.05 hPa (20 °C)
Flash Point 103 °C
Refractive Index 1.4345 (20 °C, 589 nm)
Explosion Limit 0.7 - 5.9 %(V)
Density 0.79 g/cm3 (20 °C)
pH 9.9 (0.3 g/l, H2O, 25 °C)
Ignition Point 250 °C
Hill Formula C12H27N
Chemical Formula CH3(CH2)5NH(CH2)5CH3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR