Dibenzofuran for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20408.0005 8.20408.0250
EM8.20408.0005CS 31961 USD
EM8.20408.0005 EM8.20408.0250
Dibenzofuran for synthesis, Sigma-Aldrich®
Dibenzofuran
Formula: C₁₂H₈O
MDL Number: MFCD00004968
CAS Number: 132-64-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 79 - 82 °C
Molar Mass 168.19 g/mol
Boiling Point 287 - 289 °C
Flash Point 148 °C
Density 1.3 g/cm3 (20 °C)
Synonyms Diphenylene oxide
Hill Formula C12H8O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR