1,3-Diethoxy-2-propanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1,3-Diethoxypropan-2-ol

M-Clarity™ quality level = MQ200

8.20392.0050 8.20392.0250
EM8.20392.0050CS 7378 USD
EM8.20392.0050 EM8.20392.0250
1,3-Diethoxy-2-propanol for synthesis, Sigma-Aldrich®
1,3-Diethoxy-2-propanol
Formula: C₇H₁₆O₃
MW: 148.2 g/mol
MDL Number: MFCD00026934
CAS Number: 4043-59-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 148.20 g/mol
Boiling Point 191 - 192 °C
Density 0.95 g/cm3 (20 °C)
Synonyms Glycerol α,γ-diethyl ether
Hill Formula C7H16O3
Chemical Formula C2H5OCH2CH(OH)CH2OC2H5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR