3-Chlorophenol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.20321.0050
8.20321.0250
EM8.20321.0050CS
9601
USD
EM8.20321.0050
EM8.20321.0250
3-Chlorophenol for synthesis, Sigma-Aldrich®
3-Chlorophenol
Formula:
C₆H₅ClO MW: 128.56 g/mol |
MDL Number:
MFCD00002256 CAS Number: 108-43-0 UN: 2020 ADR: 6.1,III |
Specification Test Results
Solubility | 27.7 g/l (20 °C) |
Melting Point | 31 - 34 °C |
Molar Mass | 128.55 g/mol |
Boiling Point | 216.8 °C (1013 hPa) |
Vapor Pressure | 1 hPa (44 °C) |
Flash Point | 97 °C |
Density | 1.27 g/cm3 (45 °C) |
pH | 3.5 (27.7 g/l, H2O, 20 °C) |
Ignition Point | >600 °C |
Hill Formula | C6H5ClO |
Chemical Formula | 3-(Cl)C6H4OH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...