1-Chloronaphthalene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Triisodecyl phosphite, α-Chloronaphthalene
M-Clarity™ quality level = MQ200
8.20315.0100
8203151000
EM8.20315.0100CS
837
USD
EM8.20315.0100
EM8.20315.1000
1-Chloronaphthalene for synthesis, Sigma-Aldrich®
1-Chloronaphthalene
Formula:
C₁₀H₇Cl MW: 162.62 g/mol |
MDL Number:
MFCD00003874 CAS Number: 90-13-1 UN: 3082 ADR: 6.1,III |
Specification Test Results
Solubility | 0.009 g/l (20 °C) |
Melting Point | -6 °C |
Molar Mass | 162.61 g/mol |
Boiling Point | 259 °C (1013 hPa) |
Vapor Pressure | 0.05 hPa (20 °C) |
Flash Point | 115 °C |
Density | 1.19 g/cm3 (20 °C) |
pH | 7 (H2O, 20 °C) (undiluted) |
Ignition Point | >500 °C |
Synonyms | α-Naphthyl chloride |
Hill Formula | C10H7Cl |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...