Dibutyl sulphide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Dibutylsulphide, Butyl sulphide, n-Butyl sulphide

M-Clarity™ quality level = MQ200

8.20252.0100 8.20252.0500
EM8.20252.0100CS 1704 USD
EM8.20252.0100 EM8.20252.0500
Dibutyl sulphide for synthesis, Sigma-Aldrich®
Dibutyl sulphide
Formula: C₈H₁₈S
MW: 146.3 g/mol
Boiling Pt: 189
Melting Pt: -80
Flash Pt: 62
MDL Number: MFCD00009468
CAS Number: 544-40-1
UN: 3334
Merck Index: 12,01626

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -80 °C
Molar Mass 146.29 g/mol
Boiling Point 188.7 °C (1013 hPa)
Vapor Pressure 26 hPa (80 °C)
Flash Point 62 °C
Refractive Index 1.453 (20 °C, 589 nm)
Density 0.84 g/cm3 (20 °C)
Synonyms Butyl sulfide
Hill Formula C8H18S
Chemical Formula CH3(CH2)3S(CH2)3CH3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR