Dibutyl disulfide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Dibutyldisulphide, n-Butyl disulphide
M-Clarity™ quality level = MQ200
8.20242.0025
8.20242.0100
EM8.20242.0025CS
4167
USD
EM8.20242.0025
EM8.20242.0100
Dibutyl disulfide for synthesis, Sigma-Aldrich®
Dibutyl disulphide
Formula:
C₈H₁₈S₂ MW: 178.36 g/mol |
MDL Number:
MFCD00009467 CAS Number: 629-45-8 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | -94 °C |
Molar Mass | 178.35 g/mol |
Boiling Point | 229 - 233 °C |
Flash Point | 93 °C |
Refractive Index | 1.492 (20 °C, 589 nm) |
Explosion Limit | 0.8 %(V) |
Density | 0.94 g/cm3 (20 °C) |
Synonyms | Butyl disulfide |
Hill Formula | C8H18S2 |
Chemical Formula | CH3(CH2)3SS(CH2)3CH3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...