Pyruvic acid for synthesis, Sigma-Aldrich®

Supplier: MERCK, S.A. DE C.V. SV
Danger

Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid

M-Clarity™ quality level = MQ200

8.20170.0100 8.20170.0500
EM8.20170.0100CS 3655 USD
EM8.20170.0100 EM8.20170.0500
Pyruvic acid for synthesis, Sigma-Aldrich®
Pyruvic acid
Formula: C₃H₄O₃
MW: 88.06 g/mol
MDL Number: MFCD00002585
CAS Number: 127-17-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Melting Point 12 °C
Molar Mass 88.06 g/mol
Flash Point 82 - 83 °C
Refractive Index 1.428 (20 °C, 589 nm)
Density 1.27 g/cm3 (20 °C)
pH 1.2 (90 g/l, H2O, 20 °C)
Ignition Point 305 °C
Hill Formula C3H4O3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR