3-Acetoxyindole for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
3-Indolyl Acetate, Indoxyl acetate, 3-Indoxyl acetate, 3-Indolyl acetate
M-Clarity™ quality level = MQ200
8.20706.0001
8.20706.0005
EM8.20706.0001CS
101804
USD
EM8.20706.0001
EM8.20706.0005
3-Acetoxyindole for synthesis, Sigma-Aldrich®
3-Acetoxyindole
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol |
MDL Number:
MFCD00014561 CAS Number: 608-08-2 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 127 - 129 °C |
Molar Mass | 175.19 g/mol |
Synonyms | Acetic acid 3-indolyl ester, 3-Acetoxyindole, Indoxyl acetate |
Hill Formula | C10H9NO2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...