Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulfonate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
1-Amino-8-naphthol-3,6-disulphonic acid monosodium salt, H acid monosodium salt, 8-Amino-1-naphthol-3,6-disulphonic acid monosodium salt, 4-Amino-5-hydroxy-2,7-naphthalenedisulphonic acid Monosodium Salt Hydrate, 4-Amino-5-hydroxynaphthalene-2,7-disulphonic acid monosodium salt, Sodium 4-Amino-5-hydroxy-2,7-naphthalenedisulphonate Hydrate, H acid Hydrate
M-Clarity™ quality level = MQ200
8.20078.0100
8.20078.0500
EM8.20078.0100CS
3243
USD
EM8.20078.0100
EM8.20078.0500
Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulfonate for synthesis, Sigma-Aldrich®
Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulphonate
Formula:
C₁₀H₈NNaO₇S₂ MW: 341.28 g/mol Melting Pt: >380 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150460 CAS Number: 5460-09-3 EINECS: 226-736-4 |
Specification Test Results
Assay (acidimetric) | ≥ 85.0 % |
Water (K. F.) | 8.0 - 12.0 % |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...