tert-Butylchlorodimethylsilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride

M-Clarity™ quality level = MQ200

8.18642.0025 8.18642.0100
EM8.18642.0025CS 13482 USD
EM8.18642.0025 EM8.18642.0100
tert-Butylchlorodimethylsilane for synthesis, Sigma-Aldrich®
tert-Butylchlorodimethylsilane
Formula: C₆H₁₅ClSi
MW: 150.72 g/mol
MDL Number: MFCD00000501
CAS Number: 18162-48-6
UN: 2925
ADR: 4.1 (8),II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 87 - 90 °C
Molar Mass 150.72 g/mol
Boiling Point 125 °C
Flash Point 22 °C
Synonyms tert-Butyldimethylsilyl chloride, tert-Butylchlorodimethylsilane, TBDMSCl
Hill Formula C6H15ClSi
Chemical Formula (CH3)3CSi(CH3)2Cl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR