4-Cyanobenzaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4-Formylbenzonitrile

M-Clarity™ quality level = MQ200

8.18530.0005 8.18530.0025
EM8.18530.0005CS 66030 USD
EM8.18530.0005 EM8.18530.0025
4-Cyanobenzaldehyde for synthesis, Sigma-Aldrich®
4-Cyanobenzaldehyde
Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 262
MDL Number: MFCD00003376
CAS Number: 105-07-7
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 100 - 103 °C
Molar Mass 131.14 g/mol
Boiling Point 133 °C (16 hPa)
Synonyms 4-Formylbenzonitrile
Hill Formula C8H5NO
Chemical Formula 4-(OHC)C6H4CN

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR