Copper(II) chloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Copper dichloride, Cupric chloride
M-Clarity™ quality level = MQ200
8.18247.0100
8.18247.0500
8.18247.9020
EM8.18247.0100CS
752
USD
EM8.18247.0100
EM8.18247.0500
EM8.18247.9020
Copper(II) chloride for synthesis, Sigma-Aldrich®
Copper(II) chloride
Formula:
Cl₂Cu MW: 134.45 g/mol |
MDL Number:
MFCD00010972 CAS Number: 7447-39-4 UN: 2802 ADR: 8,III |
Specification Test Results
Solubility | 620 g/l (20 °C) |
Melting Point | 498 °C decomposes |
Molar Mass | 134.45 g/mol |
Density | 3.39 g/cm3 (25 °C) |
pH | 3.5 (50 g/l, H2O, 20 °C) |
Synonyms | Copper dichloride |
Hill Formula | Cl2Cu |
Chemical Formula | CuCl2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...