9-Fluorenylmethyl chloroformate (Fmoc chloride) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
(9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl
M-Clarity™ quality level = MQ200
8.18203.0005
8.18203.0025
EM8.18203.0005CS
8899
USD
EM8.18203.0005
EM8.18203.0025
9-Fluorenylmethyl chloroformate (Fmoc chloride) for synthesis, Sigma-Aldrich®
9-Fluorenylmethyl chloroformate (Fmoc chloride)
Formula:
C₁₅H₁₁ClO₂ MW: 258.7 g/mol |
MDL Number:
MFCD00001138 CAS Number: 28920-43-6 UN: 1759 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 60 - 63 °C |
Molar Mass | 258.7 g/mol |
Synonyms | Chloroformic acid 9-fluorenylmethyl ester, FMOCCl |
Hill Formula | C15H11ClO2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...