(-)-Menthyl acetate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
L-Menthyl acetate, L-Menthol acetate, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl acetate, (R)-(-)-Menthyl acetate, (-)-Acetoxy-p-menthane, (5R)-2-Isopropyl-5-methylcyclohexyl acetate, (1R)-(−)-Menthyl acetate
M-Clarity™ quality level = MQ200
8.16147.0010
8.16147.0100
EM8.16147.0010CS
43949
USD
EM8.16147.0010
EM8.16147.0100
(-)-Menthyl acetate for synthesis, Sigma-Aldrich®
(-)-Menthyl acetate
Formula:
C₁₂H₂₂O₂ |
MDL Number:
MFCD00001482 CAS Number: 2623-23-6 |
Specification Test Results
Molar Mass | 198.3 g/mol |
Boiling Point | 229.5 °C |
Density | 0.925 g/cm3 |
Hill Formula | C12H22O2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...