N-Lauroylsarcosine sodium salt stabilised for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate
M-Clarity™ quality level = MQ200
8.14715.0100
8.14715.0500
EM8.14715.0100CS
647
USD
EM8.14715.0100
EM8.14715.0500
N-Lauroylsarcosine sodium salt stabilised for synthesis, Sigma-Aldrich®
N-Lauroylsarcosine sodium salt
Formula:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa |
MDL Number:
MFCD00042728 CAS Number: 137-16-6 UN: 2811 ADR: 6.1,II |
|
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 45 - 47 °C |
Molar Mass | 293.38 g/mol |
Bulk Density | 400 kg/m3 |
Flash Point | >150 °C |
pH | 8 (30 g/l, H2O, 20 °C) |
Synonyms | Sarcosyl, Sodium lauroylsarcosinate |
Hill Formula | C15H28NNaO3 |
Chemical Formula | CH3-(CH2)10-CO-N(CH3)-CH2-COONa |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...