Sebacoyl dichloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Sebacoyl chloride
M-Clarity™ quality level = MQ200
8.18899.0025
8.18899.0100
EM8.18899.0025CS
6096
USD
EM8.18899.0025
EM8.18899.0100
Sebacoyl dichloride for synthesis, Sigma-Aldrich®
Sebacoyl dichloride
Formula:
C₁₀H₁₆Cl₂O₂ MW: 239.14 g/mol |
MDL Number:
MFCD00000770 CAS Number: 111-19-3 UN: 3265 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | -5 °C |
Molar Mass | 239.14 g/mol |
Boiling Point | 161 °C (19 hPa) |
Flash Point | 113 °C |
Refractive Index | 1.4684 (18 °C, 589 nm) |
Density | 1.12 g/cm3 (20 °C) |
Synonyms | Sebacoyl chloride |
Hill Formula | C10H16Cl2O2 |
Chemical Formula | ClOC(CH2)8COCl |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...