Suberic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid
M-Clarity™ quality level = MQ200
8.18800.0100
8.18800.0500
EM8.18800.0100CS
1655
USD
EM8.18800.0100
EM8.18800.0500
Suberic acid for synthesis, Sigma-Aldrich®
Suberic acid
Formula:
C₈H₁₄O₄ MW: 174.2 g/mol |
MDL Number:
MFCD00004428 CAS Number: 505-48-6 |
Specification Test Results
Solubility | 1.6 g/l (20 °C) |
Melting Point | 139 - 142 °C |
Molar Mass | 174.2 g/mol |
Bulk Density | 700 kg/m3 |
Boiling Point | 229 - 231 °C (20 hPa) |
Vapor Pressure | < - 1 hPa (20 °C) |
Flash Point | 210 °C |
pH | 3.2 (1.6 g/l, H2O, 20 °C) |
Ignition Point | 430 °C |
Synonyms | Suberic acid |
Hill Formula | C8H14O4 |
Chemical Formula | HOOC(CH2)6COOH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...