(±)-2-Nonanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (±)-Heptyl methyl carbinol, (±)-2-Hydroxy nonane, (±)-Methyl heptyl carbinol

M-Clarity™ quality level = MQ200

8.18793.0025 8.18793.0100 8.18793.1000
EM8.18793.0025CS 4945 USD
EM8.18793.0025 EM8.18793.0100 EM8.18793.1000
(±)-2-Nonanol for synthesis, Sigma-Aldrich®
(±)-2-Nonanol
Formula: C₉H₂₀O
MW: 144.26 g/mol
MDL Number: MFCD00004593
CAS Number: 628-99-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -35 °C
Molar Mass 144.26 g/mol
Boiling Point 198 °C
Flash Point 96 °C
Refractive Index 1.429 (25 °C, 589 nm)
Density 0.82 g/cm3 (20 °C)
Synonyms Heptyl methyl carbinol
Hill Formula C9H20O
Chemical Formula CH3(CH2)6CH(OH)CH3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR