Melatonin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

M-Clarity™ quality level = MQ200

8.14537.0001 8.14537.0005
EM8.14537.0001CS 79113 USD
EM8.14537.0001 EM8.14537.0005
Melatonin for synthesis, Sigma-Aldrich®
Melatonin
Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
MDL Number: MFCD00005655
CAS Number: 73-31-4

Order Now

Specification Test Results

Melting Point 116 - 118 °C
Molar Mass 232.28 g/mol
Synonyms N-Acetyl-5-methoxytryptamine
Hill Formula C13H16N2O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR