Pyridinium-p-toluenesulphonate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Pyridin-1-ium 4-methylbenzenesulphonate
M-Clarity™ quality level = MQ200
8.14330.0025
8.14330.0100
EM8.14330.0025CS
9825
USD
EM8.14330.0025
EM8.14330.0100
Pyridinium-p-toluenesulphonate for synthesis, Sigma-Aldrich®
Pyridinium-p-toluenesulphonate
Formula:
C₁₂H₁₃NO₃S MW: 251.31 g/mol |
MDL Number:
MFCD00013108 CAS Number: 24057-28-1 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 112 - 119 °C (dry substance) |
Molar Mass | 251.30 g/mol |
Hill Formula | C12H13NO3S |
Chemical Formula | 4-(CH3)C6H4SO3H * C5H5N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...