Bis(2-ethylhexyl)phosphate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid
M-Clarity™ quality level = MQ200
8.14181.0250
8.14181.1000
EM8.14181.0250CS
762
USD
EM8.14181.0250
EM8.14181.1000
Bis(2-ethylhexyl)phosphate for synthesis, Sigma-Aldrich®
Bis(2-ethylhexyl)phosphate
Formula:
C₁₆H₃₅O₄P MW: 322.43 g/mol |
MDL Number:
MFCD00009492 CAS Number: 298-07-7 UN: 1902 ADR: 8 III |
Specification Test Results
Solubility | < - 1 g/l (20 °C) |
Melting Point | -50 °C |
Molar Mass | 322.42 g/mol |
Boiling Point | 48 °C (16 hPa) |
Vapor Pressure | < - 0.1 hPa (20 °C) |
Flash Point | 150 °C |
Refractive Index | 1.443 (20 °C, 589 nm) |
Density | 0.97 g/cm3 (20 °C) |
pH | 3 (< 1 g/l, H2O) |
Ignition Point | >300 °C |
Synonyms | HDEHP, Phosphoric acid bis (2-ethylhexyl) ester, Bis(2-ethylhexyl) phosphoric acid |
Hill Formula | C16H35O4P |
Chemical Formula | (CH3(CH2)3CH(C2H5)CH2O)2POOH |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...