Ruthenium(III) acetylacetonate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)
M-Clarity™ quality level = MQ200
8.14056.0001
8.14056.0005
EM8.14056.0001CS
186423
USD
EM8.14056.0001
EM8.14056.0005
Ruthenium(III) acetylacetonate for synthesis, Sigma-Aldrich®
Ruthenium(III) acetylacetonate
Formula:
C₁₅H₂₁O₆Ru MW: 398.4 g/mol |
MDL Number:
MFCD00000030 CAS Number: 14284-93-6 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 230 - 235 °C |
Molar Mass | 398.39 g/mol |
Synonyms | Tris(acetylacetonato)ruthenium(III), Tris(pentane-2,4-dionato)ruthenium(III) |
Hill Formula | C15H21O6Ru |
Chemical Formula | (CH3C(O)=CHCOCH3)3Ru |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...