Dibromomethane stabilized for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Methylenbromide
M-Clarity™ quality level = MQ200
8.10278.0100
8.10278.0250
8.10278.2500
EM8.10278.0100CS
2155
USD
EM8.10278.0100
EM8.10278.0250
EM8.10278.2500
Dibromomethane stabilized for synthesis, Sigma-Aldrich®
Dibromomethane
Formula:
Br₂CH₂ MW: 173.83 g/mol |
MDL Number:
MFCD00000168 CAS Number: 74-95-3 UN: 2664 ADR: 6.1,III |
Specification Test Results
Solubility | 11.7 g/l (15 °C) |
Melting Point | -52.7 °C |
Molar Mass | 173.83 g/mol |
Boiling Point | 95 - 97 °C (1013 hPa) |
Vapor Pressure | 44 hPa (20 °C) |
Refractive Index | 1.5419 (20 °C, 589 nm) |
Density | 2.49 g/cm3 (20 °C) |
Synonyms | Methylene bromide |
Hill Formula | CH2Br2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...