2-Undecanone ≥98% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Methyl nonyl ketone, Nonyl methyl ketone

M-Clarity™ quality level = MQ200

8.08469.0100 8.08469.0500 8.08469.9020
EM8.08469.0100CS 908 USD
EM8.08469.0100 EM8.08469.0500 EM8.08469.9020
2-Undecanone ≥98% (by GC) for synthesis, Sigma-Aldrich®
2-Undecanone
Formula: C₁₁H₂₂O
MW: 170.3 g/mol
MDL Number: MFCD00009583
CAS Number: 112-12-9
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 11 - 13 °C
Molar Mass 170.29 g/mol
Boiling Point 228 °C (1013 mbar)
Vapor Pressure 0.01 hPa (20 °C)
Flash Point 96 °C
Refractive Index 1.428 (20 °C, 589 nm)
Density 0.83 g/cm3 (20 °C)
Synonyms Methyl nonyl ketone
Hill Formula C11H22O
Chemical Formula CH3(CH2)8COCH3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR