Phenyl isothiocyanate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.07028.0100 8.07028.0250
EM8.07028.0100CS 1270 USD
EM8.07028.0100 EM8.07028.0250
Phenyl isothiocyanate for synthesis, Sigma-Aldrich®
Phenyl isothiocyanate
Formula: C₇H₅NS
MW: 135.19 g/mol
Boiling Pt: 221
Melting Pt: -21
Flash Pt: 87
MDL Number: MFCD00004798
CAS Number: 103-72-0
UN: 2206
ADR: 6.1 II
Merck Index: 12,07450

Order Now

Specification Test Results

Solubility (20 °C) insoluble,(decomposition)
Melting Point -21 °C
Molar Mass 135.18 g/mol
Boiling Point 221 °C (1013 hPa)
Vapor Pressure 10 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.6497 (20 °C, 589 nm)
Density 1.13 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Synonyms Isothiocyanic acid phenyl ester, PTC
Hill Formula C7H5NS
Chemical Formula C6H5NCS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR