Propiophenone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.07024.0250 8.07024.1000
EM8.07024.0250CS 680 USD
EM8.07024.0250 EM8.07024.1000
Propiophenone for synthesis, Sigma-Aldrich®
Propiophenone
Formula: C₆H₅C(O)CH₂CH₃
MDL Number: MFCD00009309
CAS Number: 93-55-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=796cda47787bc1a939c0285780228746da70cd03&vid=b0290578193d65d6b598ec25da82aff9335f1d45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Melting Point 18 °C
Molar Mass 134.17 g/mol
Boiling Point 214 - 218 °C (1013 hPa)
Vapor Pressure 1.99 hPa (20 °C)
Flash Point 85 °C
Refractive Index 1.5258 (20 °C, 589 nm)
Density 1.01 g/cm3 (20 °C)
Synonyms 1-Phenyl-1-propanone, Ethyl phenyl ketone
Hill Formula C9H10O
Chemical Formula C6H5COC2H5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR