Nickel(II) acetylacetonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate

M-Clarity™ quality level = MQ200

8.06715.0025 8.06715.0100 8.06715.9025
EM8.06715.0025CS 4062 USD
EM8.06715.0025 EM8.06715.0100 EM8.06715.9025
Nickel(II) acetylacetonate for synthesis, Sigma-Aldrich®
Nickel(II) acetylacetonate
Formula: C₁₀H₁₄NiO₄
MW: 256.91 g/mol
MDL Number: MFCD00000024
CAS Number: 3264-82-2

Order Now

Specification Test Results

Solubility 4.8 g/l (20 °C)
Melting Point 208 °C
Molar Mass 256.91 g/mol
Bulk Density 470 kg/m3
Boiling Point 220 - 235 °C (14 hPa)
Vapor Pressure 2.7 hPa (110 °C)
Synonyms Bis(acetylacetonato)nickel(II), Bis(2,4-pentanedionato) nickel(II)
Hill Formula C10H14NiO4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR