Sodium methoxide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.06538.0005
8.06538.0500
EM8.06538.0005CS
25466
USD
EM8.06538.0005
EM8.06538.0250
EM8.06538.0500
Sodium methoxide for synthesis, Sigma-Aldrich®
Sodium methoxide
Formula:
CH₃NaO |
MDL Number:
MFCD00012179 CAS Number: 124-41-4 UN: 1431 ADR: 4.2 (8),II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 127 °C (decomposition) |
Molar Mass | 54.02 g/mol |
Bulk Density | 500 - 600 kg/m3 |
Boiling Point | >300 °C |
Vapor Pressure | (20 °C) |
Density | 1.3 g/cm3 (20 °C) |
Ignition Point | 240 °C |
Synonyms | Sodium methoxide, Sodium methanolate |
Hill Formula | CH3NaO |
Chemical Formula | CH3ONa |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...