1-Bromonaphthalene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: α-Bromonaphthalene

M-Clarity™ quality level = MQ200

8.06210.0100 8.06210.0500 8.06210.2500
EM8.06210.0100CS 1125 USD
EM8.06210.0100 EM8.06210.0500 EM8.06210.2500
1-Bromonaphthalene for synthesis, Sigma-Aldrich®
1-Bromonaphthalene
Formula: C₁₀H₇Br
MW: 207.07 g/mol
MDL Number: MFCD00003868
CAS Number: 90-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea9a18f77a334f3a7a688e768c8efc3a557bb3ab&vid=6a108e0806e706c6cdc50ad3d7fc0539d1d6a415&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Molar Mass 207.07 g/mol
Boiling Point 281 °C (1013 hPa)
Vapor Pressure 0.013 hPa (20 °C)
Flash Point 67 °C
Refractive Index 1.6576 (20 °C, 589 nm)
Density 1.48 g/cm3 (20 °C)
Synonyms α-Naphthyl bromide
Hill Formula C10H7Br

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR