4-Methylpiperidine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.06187.0100 8.06187.0500
EM8.06187.0100CS 1162 USD
EM8.06187.0100 EM8.06187.0500
4-Methylpiperidine for synthesis, Sigma-Aldrich®
4-Methylpiperidine
Formula: C₆H₁₃N
MW: 99.18 g/mol
MDL Number: MFCD00006005
CAS Number: 626-58-4
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 4 - 5 °C
Molar Mass 99.18 g/mol
Boiling Point 121 - 127 °C
Vapor Pressure 16 hPa (20 °C)
Flash Point 13 °C
Refractive Index 1.4487 (20 °C, 589 nm)
Density 0.84 g/cm3 (20 °C)
pH 13 (100 g/l, H2O, 20 °C)
Synonyms 4-Pipecoline
Hill Formula C6H13N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR