Lithium, sticks in paraffin oil for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.05660.0025
8.05660.0100
EM8.05660.0025CS
663
USD
EM8.05660.0025
EM8.05660.0100
Lithium, sticks in paraffin oil for synthesis, Sigma-Aldrich®
Lithium
Formula:
Li |
MDL Number:
MFCD00134051 CAS Number: 7439-93-2 UN: 1415 ADR: 4.3,I |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 179 °C |
Molar Mass | 6.94 g/mol |
Boiling Point | 1347 °C |
Vapor Pressure | 1 hPa (723 °C) |
Density | 0.534 g/cm3 (20 °C) |
Hill Formula | Li |
Chemical Formula | Li |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...