Isophorone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3,5,5-Trimethyl-2-cyclohexen-1-one, 3,5,5-Trimethylcyclohex-2-enone

M-Clarity™ quality level = MQ200

8.04807.0100 8.04807.1000 8.04807.2500
EM8.04807.0100CS 476 USD
EM8.04807.0100 EM8.04807.1000 EM8.04807.2500
Isophorone for synthesis, Sigma-Aldrich®
Isophorone
Formula: C₉H₁₄O
MW: 138.21 g/mol
MDL Number: MFCD00001584
CAS Number: 78-59-1

Order Now

Specification Test Results

Solubility 14.5 g/l (20 °C)
Melting Point -8 °C
Molar Mass 138.21 g/mol
Boiling Point 210 - 216 °C (1013 hPa)
Vapor Pressure 0.33 hPa (20 °C)
Flash Point 85 °C
Refractive Index 1.4759 (20 °C, 589 nm)
Explosion Limit 0.8 - 3.8 %(V)
Density 0.92 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral,Aqueous solution
Ignition Point 460 °C
Synonyms 3,5,5-Trimethyl-2-cyclohexen-1-one
Hill Formula C9H14O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR