4-Hydroxybenzaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.04536.0100 8.04536.0250 8.04536.5000
EM8.04536.0100CS 1110 USD
EM8.04536.0100 EM8.04536.0250 EM8.04536.5000
4-Hydroxybenzaldehyde for synthesis, Sigma-Aldrich®
4-Hydroxybenzaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
MDL Number: MFCD00006939
CAS Number: 123-08-0

Order Now

Specification Test Results

Solubility 13.8 g/l (30.5 °C)
Melting Point 116 °C
Molar Mass 122.12 g/mol
Boiling Point (1013 hPa) (sublimed)
Flash Point 174 °C
Density 1.13 g/cm3 (20 °C)
Ignition Point 450 °C
Hill Formula C7H6O2
Chemical Formula 4-(OH)C6H4CHO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR