Hexylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Hexylamine, 1-Aminohexane

M-Clarity™ quality level = MQ200

8.04326.0100 8.04326.0500
EM8.04326.0100CS 657 USD
EM8.04326.0100 EM8.04326.0500
Hexylamine for synthesis, Sigma-Aldrich®
Hexylamine
Formula: H₃C(CH₂)₄CH₂NH₂
MW: 101.19 g/mol
MDL Number: MFCD00008240
CAS Number: 111-26-2
UN: 2734
ADR: 8 (3),I

Order Now

Specification Test Results

Solubility 14 g/l (20 °C)
Melting Point -19 °C
Molar Mass 101.19 g/mol
Boiling Point 130 - 132 °C (1013 hPa)
Vapor Pressure 10.6 hPa (20 °C)
Flash Point 27 °C
Refractive Index 1.418 (20 °C, 589 nm)
Explosion Limit 2.1 - 9.3 %(V)
Density 0.76 g/cm3 (20 °C)
pH 11.6 (10 g/l, H2O, 20 °C)
Ignition Point 270 °C
Viscosity kinematic 1.07 mm2/s (23 °C)
Synonyms 1-Aminohexane

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR