4'-Hydroxyacetophenone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.04311.0100 8.04311.0250
EM8.04311.0100CS 4658 USD
EM8.04311.0100 EM8.04311.0250
4'-Hydroxyacetophenone for synthesis, Sigma-Aldrich®
4'-Hydroxyacetophenone
Formula: C₈H₈O₂
MW: 136.15 g/mol
MDL Number: MFCD00002359
CAS Number: 99-93-4

Order Now

Specification Test Results

Solubility 9.9 g/l (22 °C)
Melting Point 105 - 108 °C
Molar Mass 136.15 g/mol
Flash Point 166 °C
Synonyms 4-Acetylphenol
Hill Formula C8H8O2
Chemical Formula 4-(OH)C6H4COCH3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR