1-Decanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1-Hydroxy decane, 1-Decyl alcohol, Capric alcohol, Decan-1-ol, Nonyl carbinol, Decyl alcohol

M-Clarity™ quality level = MQ200

8.03463.0100 8.03463.1000
EM8.03463.0100CS 746 USD
EM8.03463.0100 EM8.03463.1000
1-Decanol for synthesis, Sigma-Aldrich®
1-Decanol
Formula: H₃C(CH₂)₈CH₂OH
MW: 158.28 g/mol
MDL Number: MFCD00004747
CAS Number: 112-30-1
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.037 g/l (25 °C)
Melting Point 5 - 7 °C
Molar Mass 158.28 g/mol
Boiling Point 220 - 235 °C (1013 hPa)
Vapor Pressure < - 0.1 hPa (20 °C)
Flash Point 95 °C DIN 51758
Refractive Index 1.4372 (20 °C, 589 nm)
Explosion Limit 0.9 - 5.7 %(V)
Density 0.8 g/cm3 (20 °C) DIN 51757
Ignition Point 255 °C
Synonyms Capric alcohol, Decyl alcohol
Hill Formula C10H22O
Chemical Formula CH3(CH2)9OH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR