2,4-Dinitroaniline for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.03248.0005
8.03248.0250
EM8.03248.0005CS
2982
USD
EM8.03248.0005
EM8.03248.0250
2,4-Dinitroaniline for synthesis, Sigma-Aldrich®
2,4-Dinitroaniline
Formula:
C₆H₅N₃O₄ MW: 183.12 g/mol |
MDL Number:
MFCD00007151 CAS Number: 97-02-9 UN: 1596 ADR: 6.1,II |
Specification Test Results
Solubility | 1 g/l (20 °C) |
Melting Point | 177 - 180 °C |
Bulk Density | 800 kg/m3 |
Boiling Point | >300 °C decomposes |
Vapor Pressure | < - 0.001 hPa (25 °C) |
Flash Point | 224 °C |
pH | 7 (1 g/l, H2O) |
Ignition Point | >365 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...