Cinnamaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.02505.0005 8.02505.0250 8.02505.1000
EM8.02505.0005CS 4879 USD
EM8.02505.0005 EM8.02505.0250 EM8.02505.1000
Cinnamaldehyde for synthesis, Sigma-Aldrich®
Cinnamaldehyde
Formula: C₉H₈O
MDL Number: MFCD00007000
CAS Number: 104-55-2

Order Now

Specification Test Results

Solubility 1.1 g/l (20 °C)
Melting Point -8 °C
Molar Mass 132.16 g/mol
Boiling Point 127 °C (21 hPa)
Vapor Pressure < - 0.1 hPa (20 °C)
Flash Point 138 °C
Refractive Index 1.6219 (20 °C, 589 nm)
Density 1.05 g/cm3 (20 °C)
Synonyms trans-Cinnamic aldehyde, trans-3-Phenyl-2-propenal
Hill Formula C9H8O
Chemical Formula C6H5CH=CHCHO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR