1,1'-Carbonyldiimidazole for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI
M-Clarity™ quality level = MQ200
8.02301.0005
8.02301.0025
8.02301.0100
8.02301.0002
EM8.02301.0005CS
31552
USD
EM8.02301.0005
EM8.02301.0025
EM8.02301.0100
EM8.02301.0002
1,1'-Carbonyldiimidazole for synthesis, Sigma-Aldrich®
1,1'-Carbonyldiimidazole
Formula:
C₇H₆N₄O MW: 162.15 g/mol Melting Pt: 111…116 °C |
MDL Number:
MFCD00005286 CAS Number: 530-62-1 UN: 3263 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 116 - 120 °C |
Molar Mass | 162.15 g/mol |
pH | 7.7 (50 g/l, H2O, 20 °C) |
Hill Formula | C7H6N4O |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...