Benzyltrimethylammonium hydroxide 40% in methanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide

M-Clarity™ quality level = MQ200

8.01816.0100 8.01816.0250 8.01816.5000
EM8.01816.0100CS 676 USD
EM8.01816.0100 EM8.01816.0250 EM8.01816.5000
Benzyltrimethylammonium hydroxide 40% in methanol for synthesis, Sigma-Aldrich®
Benzyltrimethylammonium hydroxide
Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
MDL Number: MFCD00008281
CAS Number: 100-85-6
UN: 3286
ADR: 3 (6.1, 8),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -98 °C
Boiling Point 65 °C (1013 hPa)
Vapor Pressure 127 hPa (20 °C)
Flash Point 15 °C
Explosion Limit 5.1 - 44 %(V)
Density 0.93 g/cm3 (20 °C)
pH 12 (H2O, 20 °C) (undiluted)
Ignition Point 455 °C
Synonyms Trimethylbenzylammonium hydroxide, Triton B

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR