Benzylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: α-Aminotoluene

M-Clarity™ quality level = MQ200

8.01812.0100 8.01812.0500 8.01812.1000
EM8.01812.0100CS 686 USD
EM8.01812.0100 EM8.01812.0500 EM8.01812.1000
Benzylamine for synthesis, Sigma-Aldrich®
Benzylamine
Formula: (C₆H₅)CH₂NH₂
MW: 107.16 g/mol
Boiling Pt: 185
Melting Pt: 10
Flash Pt: 65
MDL Number: MFCD00008106
CAS Number: 100-46-9
UN: 2735
ADR: 8 II
Merck Index: 13,01127

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -42.8 °C
Molar Mass 107.15 g/mol
Boiling Point 185 °C
Vapor Pressure 1.2 hPa (20 °C)
Flash Point 72 °C
Refractive Index 1.5438 (20 °C, 589 nm)
Explosion Limit 0.7 - 8.2 %(V)
Density 0.98 g/cm3 (20 °C)
pH 11.4 (100 g/l, H2O, 20 °C)
Ignition Point 405 °C
Synonyms Phenylmethylamine
Hill Formula C7H9N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR