Ethyl benzoylacetate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Ethyl 3-oxo-3-phenylpropanoate
M-Clarity™ quality level = MQ200
8.01808.0100
8.01808.0250
EM8.01808.0100CS
796
USD
EM8.01808.0100
EM8.01808.0250
Ethyl benzoylacetate for synthesis, Sigma-Aldrich®
Ethyl benzoylacetate
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol |
MDL Number:
MFCD00009196 CAS Number: 94-02-0 |
Specification Test Results
Solubility | (20 °C) insoluble |
Molar Mass | 192.21 g/mol |
Boiling Point | 265 - 270 °C (decomposition) |
Vapor Pressure | 1 hPa (108 °C) |
Flash Point | 140 °C |
Refractive Index | 1.531 (20 °C, 589 nm) |
Density | 1.114 g/cm3 (20 °C) |
Ignition Point | 375 °C DIN 51794 |
Synonyms | Benzoylacetic acid ethyl ester, 3-Oxo-3-phenylpropionic acid ethyl ester |
Hill Formula | C11H12O3 |
Chemical Formula | C6H5COCH2COOC2H5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...