2-Aminobenzothiazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II

M-Clarity™ quality level = MQ200

8.01268.0050 8.01268.0250
EM8.01268.0050CS 7413 USD
EM8.01268.0050 EM8.01268.0250
2-Aminobenzothiazole for synthesis, Sigma-Aldrich®
2-Aminobenzothiazole
Formula: C₇H₆N₂S
MW: 150.2 g/mol
MDL Number: MFCD00005785
CAS Number: 136-95-8

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 127 - 130 °C
Molar Mass 150.2 g/mol
Synonyms 2-Benzothiazolylamine
Hill Formula C7H6N2S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR